siRNA Details
Gene Symbol: Tsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011650.3 | NP_035780.1 | 5 | 470 | ||
GenBank mRNA | AF234179.1 | - | 5 | 470 | ||
AK010737.1 | - | 5 | 468 | |||
AK150529.1 | - | 5 | 519 | |||
AK151448.1 | - | 5 | 521 | |||
AK165598.1 | - | 5 | 510 | |||
AK201202.1 | - | 2 | 113 | |||
AK219078.1 | - | 2 | 131 | |||
BC004615.1 | - | 5 | 516 | 3585366 | ||
X81464.1 | - | 5 | 470 |

More Information
Other Species Targeted

Gene Ontology Categories:
