siRNA Details
Gene Symbol: Tyr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011661.4 | NP_035791.1 | 4 | 1311 | ||
GenBank mRNA | AK014619.1 | - | 4 | 1351 | ||
AK148172.1 | - | 4 | 1334 | |||
AK148341.1 | - | 4 | 1261 | |||
AK148432.1 | - | 4 | 1290 | |||
AY526904.1 | - | 4 | 1352 | |||
BC079678.1 | - | 4 | 1261 | 30539846 | ||
D00131.1 | - | 3 | 1173 | |||
D00440.1 | - | 4 | 1311 | |||
M20234.1 | - | 4 | 1412 | |||
M24560.1 | - | 4 | 1311 | |||
X12782.1 | - | 4 | 1352 |
