siRNA Details
Gene Symbol: Tyr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011661.4 | NP_035791.1 | 4 | 1337 | ||
GenBank mRNA | AK014619.1 | - | 4 | 1377 | ||
AK148172.1 | - | 4 | 1360 | |||
AK148341.1 | - | 4 | 1287 | |||
AK148432.1 | - | 4 | 1316 | |||
AY526904.1 | - | 4 | 1378 | |||
BC079678.1 | - | 4 | 1287 | 30539846 | ||
D00131.1 | - | 3 | 1199 | |||
D00440.1 | - | 4 | 1337 | |||
M20234.1 | - | 4 | 1438 | |||
M24560.1 | - | 4 | 1337 | |||
M26729.1 | - | 4 | 1276 | |||
X12782.1 | - | 4 | 1378 |
