siRNA Details
Gene Symbol: Ube2e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009454.2 | NP_033480.1 | 3 | 601 | ||
GenBank mRNA | AF003346.1 | - | 2 | 217 | ||
AK048227.1 | - | 3 | 411 | |||
AK076011.1 | - | 3 | 408 | |||
AK168072.1 | - | 3 | 426 | |||
AK189528.1 | - | 2 | 149 | |||
BC011477.1 | - | 3 | 601 | 4195671 | ||
X92664.1 | - | 3 | 385 |
