siRNA Details
Gene Symbol: Uba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF077330.1 | - | 8 | 600 | ||
AK014433.1 | - | 9 | 643 | |||
AK032514.1 | - | 9 | 834 | |||
AK048148.1 | - | 9 | 632 | |||
AK159381.1 | - | 9 | 720 | |||
AK166655.1 | - | 8 | 590 | |||
AK168859.1 | - | 9 | 635 | |||
AY029181.1 | - | 8 | 600 | |||
BC002002.1 | - | 9 | 620 | 3590282 | ||
BC080776.1 | - | 8 | 553 |

More Information
Other Species Targeted

