siRNA Details
Gene Symbol: Uba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF077330.1 | - | 10 | 790 | ||
AK014433.1 | - | 11 | 833 | |||
AK032514.1 | - | 11 | 1024 | |||
AK048148.1 | - | 11 | 822 | |||
AK159381.1 | - | 11 | 910 | |||
AK166655.1 | - | 10 | 780 | |||
AK168859.1 | - | 11 | 825 | |||
AY029181.1 | - | 10 | 790 | |||
BC002002.1 | - | 11 | 810 | 3590282 | ||
BC080776.1 | - | 10 | 743 |
