siRNA Details
Gene Symbol: Ubr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009461.2 | NP_033487.2 | 13 | 1581 | ||
GenBank mRNA | AF061555.1 | - | 13 | 1582 | ||
AK089616.1 | - | 13 | 1481 | |||
AK132284.1 | - | 13 | 1495 | |||
AK166885.1 | - | 13 | 1479 | |||
BC090969.1 | - | 13 | 1469 | |||
BC100461.1 | - | 13 | 1488 |
