siRNA Details
Gene Symbol: Usp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013700.1 | NP_038728.1 | 8 | 1012 | ||
GenBank mRNA | AK154008.1 | - | 8 | 1001 | ||
AK182748.1 | - | 1 | 54 | |||
AK203459.1 | - | 1 | 54 | |||
BC066993.1 | - | 8 | 990 | 5689229 |
