siRNA Details
Gene Symbol: Cdh23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252635.1 | NP_001239564.1 | 26, 27 | 3517 | ||
NM_023370.3 | NP_075859.2 | 27, 28 | 3523 | |||
GenBank mRNA | AF308939.1 | - | 25, 26 | 3116 | ||
AJ888000.1 | - | 24, 25 | 3110 | |||
AJ888001.1 | - | 24, 25 | 3110 | |||
AJ888002.1 | - | 24, 25 | 3110 | |||
AY026062.1 | - | 25, 26 | 3125 |
