siRNA Details
Gene Symbol: Cdh23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252635.1 | NP_001239564.1 | 33, 35 | 4606 | ||
NM_023370.3 | NP_075859.2 | 34, 36 | 4612 | |||
GenBank mRNA | AF308939.1 | - | 32, 34 | 4205 | ||
AJ888000.1 | - | 31, 33 | 4199 | |||
AJ888001.1 | - | 31, 33 | 4199 | |||
AJ888002.1 | - | 31, 33 | 4199 | |||
AK134635.1 | - | 3, 5 | 599 | |||
AY026062.1 | - | 32, 34 | 4214 |
