siRNA Details
Gene Symbol: Golgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030035.1 | NP_084311.1 | 14 | 7516 | ||
GenBank mRNA | AK134596.1 | - | 2 | 1504 | ||
AK220188.1 | - | 2 | 3371 | |||
BC040461.1 | - | 2 | 836 | |||
BC150731.1 | - | 14 | 7493 | 9087642 |
