siRNA Details
Gene Symbol: Golgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030035.1 | NP_084311.1 | 13 | 2593 | ||
GenBank mRNA | BC150731.1 | - | 13 | 2570 | 9087642 |

More Information
Other Species Targeted

Gene Ontology Categories:
