siRNA Details
Gene Symbol: Csnka2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173861.2 | NP_776286.1 | 1 | 755 | ||
GenBank mRNA | AF463503.1 | - | 1 | 798 | ||
BC137818.1 | - | 1 | 677 | 8860837 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Csnka2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173861.2 | NP_776286.1 | 1 | 755 | ||
GenBank mRNA | AF463503.1 | - | 1 | 798 | ||
BC137818.1 | - | 1 | 677 | 8860837 |