siRNA Details
Gene Symbol: Pex6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145488.1 | NP_663463.1 | 8 | 1882 | ||
GenBank mRNA | AK143173.1 | - | 8 | 1870 | ||
AK177958.1 | - | 2 | 200 | |||
AK184357.1 | - | 2 | 200 | |||
AK184585.1 | - | 2 | 199 | |||
AK192148.1 | - | 2 | 200 | |||
AK202266.1 | - | 2 | 201 | |||
AK220525.1 | - | 8 | 1863 | |||
BC003424.1 | - | 8 | 1882 | 2609884 |
