siRNA Details
Gene Symbol: Ttc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028639.3 | NP_082915.1 | 15 | 2027 | ||
GenBank mRNA | AK040578.1 | - | 15 | 2030 | ||
AK040610.1 | - | 15 | 1998 | |||
AK050263.1 | - | 15 | 2014 | |||
AK080284.1 | - | 15 | 2006 | |||
AK153704.1 | - | 15 | 2009 | |||
AK165244.1 | - | 8 | 689 | |||
BC023773.1 | - | 11 | 985 | |||
BC042512.1 | - | 15 | 1704 | 4223801 |
