siRNA Details
Gene Symbol: Piezo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039485.4 | NP_001034574.4 | 44 | 7680 | ||
GenBank mRNA | AK031235.1 | - | 4 | 618 | ||
AK031422.1 | - | 3 | 339 | |||
BC026614.1 | - | 1 | 68 | |||
BC075677.1 | - | 5 | 771 | |||
BC172109.1 | - | 3 | 233 | |||
HQ215521.1 | - | 44 | 6760 |
