siRNA Details
Gene Symbol: Alpk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037294.1 | NP_001032371.1 | 5 | 4464 | ||
GenBank mRNA | AK132555.1 | - | 2 | 2792 | ||
AY044451.1 | - | 1 | 2186 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Alpk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037294.1 | NP_001032371.1 | 5 | 4464 | ||
GenBank mRNA | AK132555.1 | - | 2 | 2792 | ||
AY044451.1 | - | 1 | 2186 |