siRNA Details
Gene Symbol: Ipo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153774.1 | NP_722469.1 | 6, 7 | 760 | ||
GenBank mRNA | AF273672.1 | - | 6, 7 | 705 | ||
AF273673.1 | - | 6, 7 | 705 | |||
AJ309238.1 | - | 6, 7 | 699 | |||
AK003047.1 | - | 4, 5 | 372 | |||
AK036687.1 | - | 6, 7 | 710 | |||
AK173127.1 | - | 6, 7 | 700 | |||
BC056469.1 | - | 6, 7 | 727 | |||
BC079546.1 | - | 7, 8 | 871 | |||
BC098508.1 | - | 6, 7 | 710 | 30015513 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107180, XM_008769563.
Gene Ontology Categories:
