siRNA Details
Gene Symbol: Prrc2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081290.1 | NP_001074759.1 | 16 | 3670 | ||
GenBank mRNA | AK122441.1 | - | 16 | 3583 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prrc2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081290.1 | NP_001074759.1 | 16 | 3670 | ||
GenBank mRNA | AK122441.1 | - | 16 | 3583 |