siRNA Details
Gene Symbol: Slc30a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033286.2 | NP_001028458.1 | 4 | 1502 | ||
GenBank mRNA | AK053537.1 | - | 3 | 2177 | ||
AK136932.1 | - | 4 | 1310 | |||
BC054548.1 | - | 3 | 2132 | |||
BC080293.1 | - | 2 | 578 | |||
BC108419.1 | - | 4 | 890 |
