siRNA Details
Gene Symbol: Zfp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044697.2 | NP_001038162.1 | 6 | 936 | ||
NM_001044698.2 | NP_001038163.1 | 6 | 932 | |||
NM_001044700.2 | NP_001038165.1 | 6 | 888 | |||
NM_001294323.1 | NP_001281252.1 | 6 | 895 | |||
NM_178447.3 | NP_848542.1 | 7 | 973 | |||
GenBank mRNA | AF483520.1 | - | 1 | 336 | ||
AF483521.1 | - | 1 | 336 | |||
AK036776.1 | - | 7 | 963 | |||
AK131978.1 | - | 6 | 883 | |||
AK141471.1 | - | 6 | 912 | |||
BC052029.1 | - | 4 | 636 | |||
BC057171.1 | - | 6 | 902 | |||
BC089506.1 | - | 6 | 867 | 30247022 | ||
Y00850.1 | - | 4 | 768 |
