siRNA Details
Gene Symbol: Slc39a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172653.2 | NP_766241.2 | 6 | 1668 | ||
GenBank mRNA | AK049099.1 | - | 4 | 414 | ||
AK122483.1 | - | 5 | 1102 | |||
AK136117.1 | - | 5 | 1380 | |||
BC052880.1 | - | 6 | 1782 | 30045566 | ||
BC059214.1 | - | 4 | 552 | |||
BC062918.1 | - | 6 | 1668 | 6833098 |
