siRNA Details
Gene Symbol: Dolk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177648.3 | NP_808316.1 | 1 | 932 | ||
GenBank mRNA | AB073968.1 | - | 1 | 875 | ||
AK077581.1 | - | 1 | 902 | |||
AK144705.1 | - | 1 | 913 | |||
BC019543.1 | - | 1 | 877 | |||
BC026973.1 | - | 1 | 657 | 5371267 |
