siRNA Details
Gene Symbol: Dolk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177648.3 | NP_808316.1 | 1 | 1844 | ||
GenBank mRNA | AB073968.1 | - | 1 | 1787 | ||
AK077581.1 | - | 1 | 1815 | |||
AK144705.1 | - | 1 | 1825 | |||
AK214790.1 | - | 1 | 161 | |||
AK217764.1 | - | 1 | 119 | |||
BC019543.1 | - | 1 | 1789 | |||
BC026973.1 | - | 1 | 1569 | 5371267 |
