siRNA Details
Gene Symbol: Rabepk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145522.4 | NP_663497.2 | 3 | 341 | ||
GenBank mRNA | AK035343.1 | - | 3 | 221 | ||
AK153022.1 | - | 3 | 245 | |||
AK179101.1 | - | 1 | 33 | |||
AK206590.1 | - | 1 | 33 | |||
BC019800.1 | - | 3 | 217 | 5150243 |
