siRNA Details
Gene Symbol: Ppig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081086.1 | NP_001074555.1 | 9 | 886 | ||
GenBank mRNA | AK134121.1 | - | 9 | 888 | ||
AK135178.1 | - | 10 | 1018 | |||
BC048905.1 | - | 9 | 747 | |||
BC150694.1 | - | 10 | 907 | 9087605 |
