siRNA Details
Gene Symbol: Zdhhc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144887.4 | NP_659136.1 | 8 | 2054 | ||
GenBank mRNA | AK028737.1 | - | 5 | 620 | ||
AK082513.1 | - | 8 | 1979 | |||
AK145086.1 | - | 8 | 1981 | |||
AK173251.1 | - | 8 | 2009 | |||
AY894891.1 | - | 8 | 2046 | |||
BC020051.1 | - | 7 | 1024 | 3987696 | ||
BC027047.1 | - | 4 | 288 | |||
BC065155.1 | - | 8 | 1924 | 5706087 |
