siRNA Details
Gene Symbol: F830045P16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177653.3 | NP_808321.1 | 2 | 257 | ||
GenBank mRNA | AK089892.1 | - | 2 | 258 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: F830045P16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177653.3 | NP_808321.1 | 2 | 257 | ||
GenBank mRNA | AK089892.1 | - | 2 | 258 |