siRNA Details
Gene Symbol: Npepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213733.2 | NP_998898.1 | 10 | 1352 | ||
GenBank mRNA | AK129470.1 | - | 1 | 541 | ||
AK150313.1 | - | 10 | 1353 | |||
AK156136.1 | - | 10 | 1353 | |||
AK170509.1 | - | 10 | 1353 | |||
BC023239.1 | - | 9 | 1030 | |||
BC055448.1 | - | 8 | 1306 | |||
BC069933.1 | - | 10 | 1347 | 4008091 | ||
BC080707.1 | - | 10 | 1225 |
