siRNA Details
Gene Symbol: Pde4dip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039376.2 | NP_001034465.2 | 16 | 2052 | ||
NM_001289701.1 | NP_001276630.1 | 16 | 2052 | |||
NM_001289702.1 | NP_001276631.1 | 16 | 2052 | |||
NM_178080.4 | NP_835181.2 | 9 | 2560 | |||
GenBank mRNA | AK028810.1 | - | 9 | 2527 | ||
AK122286.1 | - | 9 | 2555 | |||
AK141917.1 | - | 16 | 2178 | |||
BC025653.1 | - | 9 | 2486 | |||
BC028274.1 | - | 16 | 2062 | |||
BC034266.1 | - | 9 | 1225 | |||
BC050783.1 | - | 9 | 2514 | |||
BC057350.1 | - | 9 | 2991 | |||
BC141172.1 | - | 9 | 2488 | |||
BC141185.1 | - | 16 | 2052 | 9055797 | ||
BC145504.1 | - | 16 | 2052 | 9054067 |
