siRNA Details
Gene Symbol: Pde4dip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039376.2 | NP_001034465.2 | 16, 17 | 2193 | ||
NM_001289701.1 | NP_001276630.1 | 16, 17 | 2193 | |||
NM_001289702.1 | NP_001276631.1 | 16, 17 | 2193 | |||
NM_178080.4 | NP_835181.2 | 9, 10 | 2701 | |||
GenBank mRNA | AK028810.1 | - | 9, 10 | 2668 | ||
AK122286.1 | - | 9, 10 | 2696 | |||
AK141917.1 | - | 16, 17 | 2319 | |||
BC025653.1 | - | 9, 10 | 2627 | |||
BC028274.1 | - | 16, 17 | 2203 | |||
BC034266.1 | - | 9, 10 | 1366 | |||
BC050783.1 | - | 9, 10 | 2655 | |||
BC057350.1 | - | 9, 10 | 3132 | |||
BC141172.1 | - | 9, 10 | 2629 | |||
BC141185.1 | - | 16, 17 | 2193 | 9055797 | ||
BC145504.1 | - | 16, 17 | 2193 | 9054067 |
