siRNA Details
Gene Symbol: Amigo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004293.2 | NP_001004293.1 | 2 | 1492 | ||
NM_146137.3 | NP_666249.2 | 2 | 1527 | |||
GenBank mRNA | AB167509.1 | - | 1 | 1148 | ||
AK035960.1 | - | 2 | 1423 | |||
AK173118.1 | - | 2 | 1481 | |||
AY237008.1 | - | 1 | 1041 | |||
BC010598.1 | - | 2 | 1443 | 3498778 |
