siRNA Details
Gene Symbol: Npr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173788.3 | NP_776149.1 | 16, 17 | 2776 | ||
GenBank mRNA | AK158195.1 | - | 11, 12 | 1327 | ||
BC023420.1 | - | 3, 4 | 417 | |||
BC037458.1 | - | 11, 12 | 1232 | |||
BC042470.1 | - | 16, 17 | 2776 | 5137261 | ||
BC049896.1 | - | 6, 7 | 871 |
