siRNA Details
Gene Symbol: E130308A19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015681.2 | NP_001015681.1 | 2 | 1239 | ||
NM_153158.5 | NP_694798.1 | 2 | 1239 | |||
GenBank mRNA | AK047478.1 | - | 2 | 1240 | ||
AK047601.1 | - | 2 | 1208 | |||
AK081604.1 | - | 2 | 1123 | |||
AK087509.1 | - | 2 | 1236 | |||
BC017649.1 | - | 2 | 1006 | 4924105 | ||
BC037070.1 | - | 2 | 1057 | 5004205 |
