siRNA Details
Gene Symbol: Tesk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146151.4 | NP_666263.3 | 2 | 498 | ||
GenBank mRNA | AK031814.1 | - | 2 | 491 | ||
AK047755.1 | - | 2 | 499 | |||
AK053368.1 | - | 567 | ||||
AK179384.1 | - | 1 | 73 | |||
BC019149.1 | - | 1 | 231 | 5053345 | ||
BC029766.1 | - | 2 | 476 | 4947564 |
