siRNA Details
Gene Symbol: Thrap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146153.3 | NP_666265.3 | 7 | 2207 | ||
GenBank mRNA | AK034044.1 | - | 7 | 2087 | ||
BC012655.1 | - | 4 | 893 | |||
BC040346.1 | - | 7 | 2138 | |||
BC092239.1 | - | 7 | 2123 | 30659321 |
