siRNA Details
Gene Symbol: Zfp362 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081098.1 | NP_001074567.1 | 9 | 1343 | ||
GenBank mRNA | BC039093.1 | - | 5 | 719 | ||
BC147222.1 | - | 8 | 1242 | 9056279 | ||
BC147223.1 | - | 8 | 1242 | 9056281 | ||
BC150964.1 | - | 8 | 1242 | 9087875 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191612, XM_006238923, XM_006238922.
Gene Ontology Categories:
