siRNA Details
Gene Symbol: Eif4g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256195.1 | NP_001243124.1 | 18 | 3001 | ||
NM_001256198.1 | NP_001243127.1 | 18 | 3001 | |||
NM_172703.3 | NP_766291.2 | 18 | 2905 | |||
GenBank mRNA | BC023898.1 | - | 5 | 747 | ||
BC024456.1 | - | 6 | 858 | |||
BC047531.1 | - | 7 | 1072 | |||
BC072600.1 | - | 18 | 2891 | 6410320 | ||
BC075705.1 | - | 3 | 414 | |||
BC079885.1 | - | 18 | 2955 | |||
BC082549.1 | - | 18 | 2965 |
