siRNA Details
Gene Symbol: Eif4g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256195.1 | NP_001243124.1 | 10 | 1076 | ||
NM_001256198.1 | NP_001243127.1 | 10 | 1076 | |||
NM_172703.3 | NP_766291.2 | 10 | 980 | |||
GenBank mRNA | AK019693.1 | - | 10 | 1089 | ||
AK046639.1 | - | 9 | 1005 | |||
AK054068.1 | - | 9 | 1046 | |||
BC072600.1 | - | 10 | 966 | 6410320 | ||
BC079885.1 | - | 10 | 1030 | |||
BC082549.1 | - | 10 | 1040 |

More Information
Other Species Targeted

Gene Ontology Categories:
