siRNA Details
Gene Symbol: 9330182L06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172706.3 | NP_766294.2 | 10 | 1382 | ||
GenBank mRNA | AK034358.1 | - | 10 | 1414 | ||
AK036245.1 | - | 10 | 1458 | |||
AK045969.1 | - | 10 | 1416 | |||
AK048738.1 | - | 10 | 1368 | |||
AK079644.1 | - | 9 | 1313 | |||
AK082474.1 | - | 9 | 1327 | |||
AK133610.1 | - | 10 | 1383 | |||
AK134808.1 | - | 10 | 1444 | |||
AK161902.1 | - | 10 | 1443 | |||
BC132290.1 | - | 10 | 1358 | 40130567 | ||
BC138012.1 | - | 10 | 1358 | 8861031 |
