siRNA Details
Gene Symbol: Arhgap24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286468.1 | NP_001273397.1 | 4 | 554 | ||
NM_029270.2 | NP_083546.2 | 6 | 1156 | |||
NM_146161.3 | NP_666273.1 | 5 | 962 | |||
GenBank mRNA | AK039617.1 | - | 3 | 1445 | ||
AK080986.1 | - | 6 | 1156 | |||
AK086270.1 | - | 1 | 650 | |||
AK134817.1 | - | 4 | 780 | |||
BC023344.1 | - | 4 | 568 | 5064885 | ||
BC025502.1 | - | 5 | 811 | 5321985 | ||
BC027070.1 | - | 3 | 400 | 5358648 |
