siRNA Details
Gene Symbol: Lrrc8d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122768.1 | NP_001116240.1 | 3 | 2612 | ||
NM_178701.3 | NP_848816.3 | 3 | 2602 | |||
GenBank mRNA | AK046249.1 | - | 3 | 2611 | ||
AK050400.1 | - | 3 | 2572 | |||
AK080722.1 | - | 3 | 2546 | |||
AK137459.1 | - | 3 | 2613 | |||
AK167985.1 | - | 2 | 2320 | |||
AK213038.1 | - | 40 | ||||
AK213350.1 | - | 1 | 166 | |||
BC014756.1 | - | 1 | 1636 | |||
BC037717.1 | - | 3 | 2583 | 4239389 |
