siRNA Details
Gene Symbol: Ssh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198109.4 | NP_932777.2 | 12 | 1083 | ||
GenBank mRNA | AB099287.1 | - | 12 | 1118 | ||
AK089308.1 | - | 12 | 1087 | |||
AK155557.1 | - | 12 | 1108 | |||
AK165380.1 | - | 11 | 1048 | |||
AK170787.1 | - | 11 | 1008 | |||
AK171210.1 | - | 12 | 1132 | |||
AK171556.1 | - | 12 | 1098 | |||
AK173144.1 | - | 11 | 1039 | |||
BC046529.1 | - | 6 | 565 |
