siRNA Details
Gene Symbol: Ssh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198109.4 | NP_932777.2 | 14 | 1637 | ||
GenBank mRNA | AB099287.1 | - | 14 | 1672 | ||
AK089308.1 | - | 14 | 1642 | |||
AK155557.1 | - | 14 | 1662 | |||
AK165380.1 | - | 13 | 1603 | |||
AK170787.1 | - | 13 | 1562 | |||
AK171210.1 | - | 14 | 1686 | |||
AK171556.1 | - | 14 | 1652 | |||
AK173144.1 | - | 13 | 1593 | |||
BC046529.1 | - | 8 | 1119 |
