siRNA Details
Gene Symbol: Tnrc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122730.2 | NP_001116202.2 | 4 | 1918 | ||
NM_178242.2 | NP_839973.1 | 4 | 1918 | |||
GenBank mRNA | AK141849.1 | - | 1 | 1009 | ||
BC049818.1 | - | 4 | 1918 | 6511384 |
