siRNA Details
Gene Symbol: Wnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185020.1 | NP_001171949.1 | 3 | 2019 | ||
NM_001185021.1 | NP_001171950.1 | 3 | 2019 | |||
NM_001199083.1 | NP_001186012.1 | 3 | 2019 | |||
NM_001199084.1 | NP_001186013.1 | 3 | 2019 | |||
NM_198703.3 | NP_941992.2 | 3 | 2019 | |||
GenBank mRNA | AK133738.1 | - | 3 | 2021 | ||
AK172935.1 | - | 4 | 1256 | |||
AY309076.1 | - | 3 | 1143 | |||
AY455867.1 | - | 3 | 1161 | |||
BC055788.1 | - | 4 | 1880 | |||
BC138445.1 | - | 3 | 1023 | 8861465 | ||
BC145282.1 | - | 3 | 1023 | 9053836 | ||
BC171955.1 | - | 3 | 1023 | 9054241 |
