siRNA Details
Gene Symbol: Wnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185020.1 | NP_001171949.1 | 26 | 7328 | ||
NM_001185021.1 | NP_001171950.1 | 26 | 7529 | |||
NM_001199083.1 | NP_001186012.1 | 28 | 8822 | |||
NM_001199084.1 | NP_001186013.1 | 28 | 8849 | |||
NM_198703.3 | NP_941992.2 | 28 | 8075 | |||
GenBank mRNA | AY309076.1 | - | 28 | 7199 | ||
AY455867.1 | - | 26 | 6479 | |||
BC054836.1 | - | 2 | 451 | |||
BC056761.1 | - | 10 | 2561 | |||
BC138445.1 | - | 26 | 6332 | 8861465 | ||
BC145282.1 | - | 26 | 6533 | 9053836 | ||
BC171955.1 | - | 26 | 6332 | 9054241 |
