siRNA Details
Gene Symbol: BC048546 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001179.3 | NP_001001179.2 | 14 | 1736 | ||
GenBank mRNA | AK138836.1 | - | 14 | 1737 | ||
AK161068.1 | - | 1 | 86 | |||
AK161184.1 | - | 14 | 1739 | |||
BC040781.1 | - | 2 | 188 |
