siRNA Details
Gene Symbol: BC048546 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001179.3 | NP_001001179.2 | 22 | 2835 | ||
GenBank mRNA | AK138836.1 | - | 22 | 2836 | ||
AK161068.1 | - | 9 | 1185 | |||
AK161184.1 | - | 22 | 2839 | |||
BC040781.1 | - | 10 | 1287 |
