siRNA Details
Gene Symbol: Fus | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139149.2 | NP_631888.1 | 5 | 473 | ||
GenBank mRNA | AF224264.1 | - | 5 | 398 | ||
AK032815.1 | - | 5 | 462 | |||
AK038226.1 | - | 5 | 459 | |||
AK040203.1 | - | 2 | 247 | |||
AK086587.1 | - | 2 | 238 | |||
AK132141.1 | - | 466 | ||||
AK134491.1 | - | 5 | 466 | |||
AK139960.1 | - | 5 | 472 | |||
AK145582.1 | - | 5 | 463 | |||
AK146198.1 | - | 5 | 466 | |||
AK149118.1 | - | 5 | 465 | |||
AK172554.1 | - | 5 | 458 | |||
BC011078.1 | - | 5 | 456 | |||
BC040827.1 | - | 5 | 428 | 4503820 | ||
BC058247.1 | - | 5 | 448 | 6314291 |
